accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3SHNH3C

CAS RN 886851-27-0

IUPAC Name: methyl[(4-methylthiophen-2-yl)methyl]amine
Synonyms: methyl[(4-methylthiophen-2-yl)methyl]amine
Molecular Weight (g/mol): 141.23
Molecular Formula: C7H11NS
InChi Key: JICZWIQRPDVYNI-UHFFFAOYSA-N
SMILES: CNCC1=CC(C)=CS1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results