Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

N-Methyl(4-methylthien-2-yl)methylamine 97+%, Thermo Scientific™

Catalog No. CC30646DA
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC30646DA 1 g Amber Glass Bottle
CC30646CB 250 mg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC30646DA Supplier Thermo Scientific Chemicals Supplier No. CC30646DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 886851-27-0
Molecular Formula C7H11NS
Molecular Weight (g/mol) 141.23
MDL Number MFCD08435851
InChI Key JICZWIQRPDVYNI-UHFFFAOYSA-N
Synonym n-methyl 4-methylthien-2-yl methylamine, methyl 4-methylthiophen-2-yl methyl amine, 2-thiophenemethanamine,n,4-dimethyl, 2-thiophenemethanamine, n,4-dimethyl, methyl 4-methyl 2-thienyl methyl amine, n-methyl-1-4-methylthiophen-2-yl methanamine
PubChem CID 18525716
IUPAC Name N-methyl-1-(4-methylthiophen-2-yl)methanamine
SMILES CNCC1=CC(C)=CS1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 141.24
Percent Purity 97%
Chemical Name or Material N-Methyl(4-methylthien-2-yl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.