accessibility menu, dialog, popup

UserName

Viewing profile as user:
H3COCH3OCH3

CAS RN 88702-69-6

IUPAC Name: 1,4-dimethoxy-2-octadecylbenzene
Synonyms: 1,4-dimethoxy-2-octadecylbenzene
Molecular Weight (g/mol): 390.65
Molecular Formula: C26H46O2
InChi Key: ZBJNNOTWURHXIS-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCCCC1=C(OC)C=CC(OC)=C1
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results