Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Octadecyl-1,4-dimethoxybenzene, TCI America™
SDP

Catalog No. O02175G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02175G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. O02175G Supplier TCI America Supplier No. O02175G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 88702-69-6
Molecular Formula C26H46O2
Molecular Weight (g/mol) 390.652
MDL Number MFCD00142955
InChI Key ZBJNNOTWURHXIS-UHFFFAOYSA-N
Synonym Octadecylhydroxyquinone Dimethyl Ether
PubChem CID 13173901
IUPAC Name 1,4-dimethoxy-2-octadecylbenzene
SMILES CCCCCCCCCCCCCCCCCCC1=C(C=CC(=C1)OC)OC

Specifications

Melting Point 54°C
Color White
Quantity 5 g
Formula Weight 390.65
Physical Form Crystalline Powder
Chemical Name or Material 2-Octadecyl-1,4-dimethoxybenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.