accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 887919-35-9

IUPAC Name: bis(4-(di-tert-butylphosphanyl)-N,N-dimethylaniline); dichloropalladium
Synonyms: bis(4-(di-tert-butylphosphanyl)-N,N-dimethylaniline); palladium chloride
Molecular Weight (g/mol): 708.08
Molecular Formula: C32H56Cl2N2P2Pd
InChi Key: DWOZNANUEDYIOF-UHFFFAOYSA-L
SMILES: Cl[Pd]Cl.CN(C)C1=CC=C(C=C1)P(C(C)(C)C)C(C)(C)C.CN(C)C1=CC=C(C=C1)P(C(C)(C)C)C(C)(C)C
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (3)
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)