Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Bis(Di-Tert-Butyl(4-Dimethylaminophenyl)Phosphine)Dichloropalladium(II)

Catalog No. 6787405G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
1 g
5 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
67-874-05G 5 g
67-874-01G 1 g
67-874-050G 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Warning
Catalog No. 67-874-05G Supplier Sigma Aldrich Supplier No. 6787405G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Higly active, air-stable catalyst for Suzuki-Miyaura cross-coupling with aryl halides including 5- and 6-membered heteroaryl chlorides.

Specifications

CAS 887919-35-9
Molecular Formula C32H56Cl2N2P2Pd
Linear Formula C32H56Cl2N2P2Pd
MDL Number MFCD09265123
Quantity 5 g
Synonym (A-taPhos)2PdCl2; Pd(amphos)Cl2
Molecular Weight (g/mol) 708.07 g/mol
Biological Activity Respiratory System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.