accessibility menu, dialog, popup

UserName

Viewing profile as user:
NNO

CAS RN 887922-90-9

IUPAC Name: 4-[(1H-pyrazol-1-yl)methyl]benzaldehyde
Synonyms: 4-(pyrazol-1-ylmethyl)benzaldehyde
Molecular Weight (g/mol): 186.21
Molecular Formula: C11H10N2O
InChi Key: FAURNROAEFQBHX-UHFFFAOYSA-N
SMILES: O=CC1=CC=C(CN2C=CC=N2)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results