Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(1H-pyrazol-1-ylmethyl)benzaldehyde, Thermo Scientific™

Catalog No. CC56004DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC56004DE 5 g
CC56004CB 250 mg
CC56004DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC56004DE Supplier Thermo Scientific Chemicals Supplier No. CC56004DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 887922-90-9
Molecular Formula C11H10N2O
Molecular Weight (g/mol) 186.21
MDL Number MFCD08059835
InChI Key FAURNROAEFQBHX-UHFFFAOYSA-N
Synonym 4-1h-pyrazol-1-ylmethyl benzaldehyde, 4-pyrazol-1-ylmethyl-benzaldehyde, 4-1h-pyrazol-1-yl methyl benzaldehyde, 4-pyrazol-1-ylmethyl benzaldehyde, 4-pyrazolylmethyl benzaldehyde, 4-1h-pyrazol-1-ylmethyl-benzaldehyde, benzaldehyde,4-1h-pyrazol-1-ylmethyl
PubChem CID 18525876
IUPAC Name 4-(pyrazol-1-ylmethyl)benzaldehyde
SMILES O=CC1=CC=C(CN2C=CC=N2)C=C1

Specifications

Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 5 g
Formula Weight 186.21
Percent Purity 97%
Chemical Name or Material 4-(1H-Pyrazol-1-ylmethyl)benzaldehyde
Hazard Category GHS Signal Word: Warning
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.