accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 88805-76-9

IUPAC Name: 5-iodo-1H-indazol-3-amine
Synonyms: 5-iodo-1H-indazol-3-amine
Molecular Weight (g/mol): 259.05
Molecular Formula: C7H6IN3
InChi Key: GBUVSJWTWOWRSL-UHFFFAOYSA-N
SMILES: NC1=NNC2=CC=C(I)C=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results