Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

3-Amino-5-iodo-1H-indazole, 95%, Thermo Scientific™

Catalog No. AC435800010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC435800010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC435800010 Supplier Thermo Scientific Chemicals Supplier No. 435800010
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 88805-76-9
Molecular Formula C7H6IN3
Molecular Weight (g/mol) 259.05
MDL Number MFCD09261138
InChI Key GBUVSJWTWOWRSL-UHFFFAOYSA-N
Synonym 3-amino-5-iodo-1h-indazole, 1h-indazol-3-amine,5-iodo, 1h-indazol-3-amine, 5-iodo, 3-amino-5-iodoindazole, acmc-20f1u4, 5-iodo-1h-indazole-3-ylamine
PubChem CID 20135868
IUPAC Name 5-iodo-1H-indazol-3-amine
SMILES NC1=NNC2=CC=C(I)C=C12

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 182°C to 188°C
Color White to Brown
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 259.05
Percent Purity 95%
Physical Form Crystalline Powder
Chemical Name or Material 3-Amino-5-iodo-1H-indazole
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.