accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONHOHN

CAS RN 89-24-7

IUPAC Name: 5-phenylimidazolidine-2,4-dione
Synonyms: phenylhydantoin 5-phenyl-2,4-imidazolidione
Molecular Weight (g/mol): 176.18
Molecular Formula: C9H8N2O2
InChi Key: NXQJDVBMMRCKQG-UHFFFAOYNA-N
SMILES: O=C1NC(C(=O)N1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results