Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Phenylhydantoin 99.0+%, TCI America™
SDP

Catalog No. P066625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P066625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P066625G Supplier TCI America Supplier No. P066625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 89-24-7
Molecular Formula C9H8N2O2
Molecular Weight (g/mol) 176.175
MDL Number MFCD00037881
InChI Key NXQJDVBMMRCKQG-UHFFFAOYSA-N
PubChem CID 1002
IUPAC Name 5-phenylimidazolidine-2,4-dione
SMILES C1=CC=C(C=C1)C2C(=O)NC(=O)N2

Specifications

Melting Point 182°C
Color White
Quantity 25 g
Formula Weight 176.18
Percent Purity ≥99.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Phenylhydantoin
EINECSNumber (5)-3495
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.