accessibility menu, dialog, popup

UserName

Viewing profile as user:
ONOClClCl

CAS RN 89-69-0

IUPAC Name: 1,2,4-trichloro-5-nitrobenzene
Synonyms: 2,4,5-trichloronitrobenzene
Molecular Weight (g/mol): 226.44
Molecular Formula: C6H2Cl3NO2
InChi Key: IBRBMZRLVYKVRF-UHFFFAOYSA-N
SMILES: [O-][N+](=O)C1=CC(Cl)=C(Cl)C=C1Cl
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results