Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2,4,5-Trichloronitrobenzene 99.0+%, TCI America™
SDP

Catalog No. T038725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T038725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T038725G Supplier TCI America Supplier No. T038725G
Add to Cart
Add to Cart

Specifications

CAS 89-69-0
Melting Point 58°C
Color Yellow
Boiling Point 150°C
Molecular Formula C6H2Cl3NO2
Quantity 25 g
MDL Number MFCD00007072
Synonym 2,4,5-trichloronitrobenzene, benzene, 1,2,4-trichloro-5-nitro, 1-nitro-2,4,5-trichlorobenzene, 3,4,6-trichloronitrobenzene, 5-nitro-1,2,4-trichlorobenzene, 2,4,5-trichloro-1-nitrobenzene, unii-0yfy8pzd2m, 0yfy8pzd2m, 2,5-trichloronitrobenzene, wln: wnr bg dg eg
InChI Key IBRBMZRLVYKVRF-UHFFFAOYSA-N
SMILES [O-][N+](=O)C1=CC(Cl)=C(Cl)C=C1Cl
IUPAC Name 1,2,4-trichloro-5-nitrobenzene
Molecular Weight (g/mol) 226.44
PubChem CID 6983
Formula Weight 226.44
Percent Purity ≥99.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,4,5-Trichloronitrobenzene
Show More Show Less
EINECSNumber (3)-0459
RTECSNumber DC2275000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.