CAS RN 89-78-1
(+/-)-Menthol 98.0+%, TCI America™
CAS: 89-78-1 Molecular Formula: C10H20O Molecular Weight (g/mol): 156.269 MDL Number: MFCD00001484 InChI Key: NOOLISFMXDJSKH-IVZWLZJFSA-N Synonym: menthol,dl-menthol,2-isopropyl-5-methylcyclohexanol,menthyl alcohol,cyclohexanol, 5-methyl-2-1-methylethyl,p-menthan-3-ol,racementhol,caswell no. 540,3-p-menthol,+-neo-menthol PubChem CID: 6566020 IUPAC Name: (1R,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol SMILES: CC1CCC(C(C1)O)C(C)C
DL-Menthol, Crystal, USP, 98-102%, Spectrum™ Chemical
CAS: 89-78-1 Molecular Formula: C10H20O Molecular Weight (g/mol): 156.27 InChI Key: NOOLISFMXDJSKH-KXUCPTDWSA-N IUPAC Name: (1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol SMILES: CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O
DL-Menthol, 99%
CAS: 89-78-1 Molecular Formula: C10H20O Molecular Weight (g/mol): 156.27 MDL Number: MFCD00001484 InChI Key: NOOLISFMXDJSKH-IVZWLZJFSA-N Synonym: menthol,dl-menthol,2-isopropyl-5-methylcyclohexanol,menthyl alcohol,cyclohexanol, 5-methyl-2-1-methylethyl,p-menthan-3-ol,racementhol,caswell no. 540,3-p-menthol,+-neo-menthol PubChem CID: 6566020 IUPAC Name: (1R,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol SMILES: CC1CCC(C(C1)O)C(C)C
DL-Menthol, 98+%
CAS: 89-78-1 Molecular Formula: C10H20O Molecular Weight (g/mol): 156.269 MDL Number: MFCD00001484 InChI Key: NOOLISFMXDJSKH-IVZWLZJFSA-N Synonym: menthol,dl-menthol,2-isopropyl-5-methylcyclohexanol,menthyl alcohol,cyclohexanol, 5-methyl-2-1-methylethyl,p-menthan-3-ol,racementhol,caswell no. 540,3-p-menthol,+-neo-menthol PubChem CID: 6566020 IUPAC Name: (1R,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol SMILES: CC1CCC(C(C1)O)C(C)C
DL-Menthol, Crystal, FCC, Spectrum™ Chemical
CAS: 89-78-1 Molecular Formula: C10H20O Molecular Weight (g/mol): 156.27 InChI Key: NOOLISFMXDJSKH-KXUCPTDWSA-N IUPAC Name: (1R,2S,5R)-5-methyl-2-(propan-2-yl)cyclohexan-1-ol SMILES: CC(C)[C@@H]1CC[C@@H](C)C[C@H]1O
Sigma Aldrich (+/-)-Menthol
MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.
Fisher Science Education™ Menthol
CAS: 89-78-1 Molecular Formula: C10H20O Molecular Weight (g/mol): 156.269 InChI Key: NOOLISFMXDJSKH-IVZWLZJFSA-N Synonym: menthol,dl-menthol,2-isopropyl-5-methylcyclohexanol,menthyl alcohol,cyclohexanol, 5-methyl-2-1-methylethyl,p-menthan-3-ol,racementhol,caswell no. 540,3-p-menthol,+-neo-menthol PubChem CID: 6566020 IUPAC Name: (1R,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol SMILES: CC1CCC(C(C1)O)C(C)C