Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Menthol
SCI_ED

Catalog No. S25751A Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Solid
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
S25751A 100 g Solid
S25751 25 g Solid
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. S25751A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 89-78-1
Molecular Formula C10H20O
Molecular Weight (g/mol) 156.269
InChI Key NOOLISFMXDJSKH-IVZWLZJFSA-N
Synonym menthol, dl-menthol, 2-isopropyl-5-methylcyclohexanol, menthyl alcohol, cyclohexanol, 5-methyl-2-1-methylethyl, p-menthan-3-ol, racementhol, caswell no. 540, 3-p-menthol, +-neo-menthol
PubChem CID 6566020
IUPAC Name (1R,2R,5S)-5-methyl-2-propan-2-ylcyclohexan-1-ol
SMILES CC1CCC(C(C1)O)C(C)C

Specifications

Color White
Density 0.890g/cmsuperscript_3;
Assay Percent Range 99% w/v
Packaging Solid
Linear Formula C10H20O
Quantity 100 g
Formula Weight 156.26
Chemical Name or Material Menthol
Safety handling Check Alert storage code Red
ShelfLife 5 years
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.