accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 89-99-6

IUPAC Name: 1-(2-fluorophenyl)methanamine
Synonyms: 1-(2-fluorophenyl)methanamine
Molecular Weight (g/mol): 125.15
Molecular Formula: C7H8FN
InChi Key: LRFWYBZWRQWZIM-UHFFFAOYSA-N
SMILES: NCC1=CC=CC=C1F
Boiling Point 
  • (3)
  • (2)
  • (3)
Molecular Weight (g/mol) 
  • (5)
  • (3)
Percent Purity 
  • (2)
  • (3)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (2)
  • (6)
  • (3)
  • (2)
  • (3)
  • (5)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (5)