accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3CH3SiCH3CH3OBr

CAS RN 89031-84-5

IUPAC Name: (3-bromopropoxy)(tert-butyl)dimethylsilane
Synonyms: (3-bromopropoxy)(tert-butyl)dimethylsilane
Molecular Weight (g/mol): 253.26
Molecular Formula: C9H21BrOSi
InChi Key: QGMROEZDWJTIDW-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](C)(C)OCCCBr
Boiling Point 
  • (2)
  • (2)
  • (2)
Molecular Weight (g/mol) 
  • (2)
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (4)
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (4)
  • (1)
  • (2)
  • (1)
  • (2)
  • (4)