Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich (3-Bromopropoxy)-tert-butyldimethylsilane

Catalog No. 4290665ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
42-906-65ML 5 mL
42-906-625ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 42-906-65ML Supplier Sigma Aldrich Supplier No. 4290665ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • (3-bromopropoxy)-tert-butyldimethylsilane is a bromo silyl ether.(3-bromopropoxy)-tert-butyldimethylsilane may be used to introduce propanol functionality to many pharmaceuticals.it may be used as an alkylating agent in the synthesis of the following:
  • N-[2-[n-[3-(tert-butyldimethylsilyloxy)propyl]-n-ethylamino]ethyl]phthalimide
  • O-(3-tert-butyldimethylsilyloxypropyl)-n-(tert-butoxycarbonyl)-L-tyrosine methyl ester
  • Tert-butyldimethyl-[3-(3-methyl-2-nitrophenoxy)propoxy]silane

Specifications

CAS 89031-84-5
Density 1.093 g/mL (at 25°C (literature))
Boiling Point 182°C (lit.)
Molecular Formula C9H21BrOSi
Refractive Index n20/D 1.451 (literature)
Linear Formula Br(CH2)3 OSi(CH3)2 C(CH3)3
MDL Number MFCD00216589
Quantity 5 mL
Molecular Weight (g/mol) 253.25
Percent Purity 97%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.