accessibility menu, dialog, popup

UserName

Viewing profile as user:
BOOH3CCH3H3CH3CFOCH3F

CAS RN 890839-37-9

IUPAC Name: 2-(3,5-difluoro-4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Synonyms: 2-(3,5-difluoro-4-methoxyphenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
Molecular Weight (g/mol): 270.08
Molecular Formula: C13H17BF2O3
InChi Key: LVDNSBZUAAXPPZ-UHFFFAOYSA-N
SMILES: COC1=C(F)C=C(C=C1F)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results