accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 89114-74-9

IUPAC Name: N,N-diphenyl-4-(2-phenylethenyl)aniline
Synonyms: N,N-diphenyl-4-(2-phenylethenyl)aniline
Molecular Weight (g/mol): 347.46
Molecular Formula: C26H21N
InChi Key: DXYYLUGHPCHMRQ-UHFFFAOYSA-N
SMILES: C(=CC1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results