Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-Styryltriphenylamine 98.0+%, TCI America™
SDP

Catalog No. S09241G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S09241G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S09241G Supplier TCI America Supplier No. S09241G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 89114-74-9
Molecular Formula C26H21N
Molecular Weight (g/mol) 347.46
MDL Number MFCD16619204
InChI Key DXYYLUGHPCHMRQ-UHFFFAOYSA-N
Synonym 4-Diphenylaminostilbene
PubChem CID 98007462
IUPAC Name N,N-diphenyl-4-(2-phenylethenyl)aniline
SMILES C(=CC1=CC=C(C=C1)N(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 153°C
Quantity 1 g
Formula Weight 347.46
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Styryltriphenylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.