accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 89115-20-8

IUPAC Name: 4-[bis(4-methoxyphenyl)amino]benzaldehyde
Synonyms: 4-[bis(4-methoxyphenyl)amino]benzaldehyde
Molecular Weight (g/mol): 333.39
Molecular Formula: C21H19NO3
InChi Key: CTRXZOLNEVJBDX-UHFFFAOYSA-N
SMILES: COC1=CC=C(C=C1)N(C1=CC=C(OC)C=C1)C1=CC=C(C=O)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results