Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-[Bis(4-methoxyphenyl)amino]benzaldehyde 98.0+%, TCI America™
SDP

Catalog No. B49051G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49051G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B49051G Supplier TCI America Supplier No. B49051G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 89115-20-8
Molecular Formula C21H19NO3
Molecular Weight (g/mol) 333.387
MDL Number MFCD16659083
InChI Key CTRXZOLNEVJBDX-UHFFFAOYSA-N
Synonym 4-Formyl-4′C,4′C′C-dimethoxytriphenylamine
PubChem CID 11256070
IUPAC Name 4-(4-methoxy-N-(4-methoxyphenyl)anilino)benzaldehyde
SMILES COC1=CC=C(C=C1)N(C2=CC=C(C=C2)C=O)C3=CC=C(C=C3)OC

Specifications

Melting Point 101°C
Color Yellow
Quantity 1 g
Formula Weight 333.39
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 4-[Bis(4-methoxyphenyl)amino]benzaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.