accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 89131-11-3

IUPAC Name: 2-(4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzenesulfonamido)-3-methylbutanoic acid
Synonyms: 2-(4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzenesulfonamido)-3-methylbutanoic acid
Molecular Weight (g/mol): 404.49
Molecular Formula: C19H24N4O4S
InChi Key: AJSKPBGSGLIRQK-UHFFFAOYNA-N
SMILES: CC(C)C(NS(=O)(=O)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)N(C)C)C(O)=O
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results