Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Dabsyl-L-valine 98.0+%, TCI America™
SDP

Catalog No. D1460100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D1460100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D1460100MG Supplier TCI America Supplier No. D1460100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 89131-11-3
Molecular Formula C19H24N4O4S
Molecular Weight (g/mol) 404.485
MDL Number MFCD00059366
InChI Key AJSKPBGSGLIRQK-SFHVURJKSA-N
Synonym 4-Dimethylaminoazobenzene-4′C-sulfonyl-L-valine, Dbs-Val-OH
PubChem CID 14048723
IUPAC Name (2S)-2-[[4-[[4-(dimethylamino)phenyl]diazenyl]phenyl]sulfonylamino]-3-methylbutanoic acid
SMILES CC(C)C(C(=O)O)NS(=O)(=O)C1=CC=C(C=C1)N=NC2=CC=C(C=C2)N(C)C

Specifications

Quantity 100 mg
Formula Weight 404.49
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Dabsyl-L-valine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.