accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OONOH

CAS RN 89151-44-0

IUPAC Name: tert-butyl 4-(2-hydroxyethyl)piperidine-1-carboxylate
Synonyms: tert-butyl 4-(2-hydroxyethyl)piperidine-1-carboxylate
Molecular Weight (g/mol): 229.32
Molecular Formula: C12H23NO3
InChi Key: YBNJZIDYXCGAPX-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)N1CCC(CCO)CC1
Boiling Point 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results