accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3H3CCH3OOHNSH2N

CAS RN 89226-13-1

IUPAC Name: tert-butyl N-(carbamothioylmethyl)carbamate
Synonyms: tert-butyl N-(carbamothioylmethyl)carbamate
Molecular Weight (g/mol): 190.26
Molecular Formula: C7H14N2O2S
InChi Key: AGBIUUFZUPNDTM-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC(N)=S

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results