Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

tert-Butyl 2-amino-2-thioxoethylcarbamate, 90%, Thermo Scientific™

Catalog No. MO07925EA
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MO07925EA 10 g Amber Glass Bottle
MO07925DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MO07925EA Supplier Thermo Scientific Chemicals Supplier No. MO07925EA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 89226-13-1
Molecular Formula C7H14N2O2S
Molecular Weight (g/mol) 190.261
MDL Number MFCD09025922
InChI Key AGBIUUFZUPNDTM-UHFFFAOYSA-N
Synonym tert-butyl 2-amino-2-thioxoethyl carbamate, tert-butyl 2-amino-2-thioxoethylcarbamate, tert-butyl n-carbamothioylmethyl carbamate, tert-butyl n-2-amino-2-sulfanylideneethyl carbamate, carbamic acid, 2-amino-2-thioxoethyl-, 1,1-dimethylethyl ester, carbamic acid,n-2-amino-2-thioxoethyl-, 1,1-dimethylethyl ester, acmc-20ljf8, n-tert-butoxycarbonylglycinethioamide, tert-butoxycarbonylamino acetothioamide, n-t-butoxycarbonylamino acetothioamide
PubChem CID 5324304
IUPAC Name tert-butyl N-(2-amino-2-sulfanylideneethyl)carbamate
SMILES CC(C)(C)OC(=O)NCC(=S)N

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 190.27
Percent Purity 90%
Chemical Name or Material Tert-butyl 2-amino-2-thioxoethylcarbamate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if swallowed.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.