accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 892501-94-9

IUPAC Name: 4-(4-isocyanatophenoxy)oxane
Synonyms: 4-(4-isocyanatophenoxy)oxane
Molecular Weight (g/mol): 219.24
Molecular Formula: C12H13NO3
InChi Key: ZGNHUEWFJQLPDN-UHFFFAOYSA-N
SMILES: O=C=NC1=CC=C(OC2CCOCC2)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results