CAS RN 892502-08-8
IUPAC Name:
methyl({4-[(1H-pyrazol-1-yl)methyl]phenyl}methyl)amine
Synonyms:
methyl({[4-(pyrazol-1-ylmethyl)phenyl]methyl})amine
Molecular Weight (g/mol):
201.27
Molecular Formula:
C12H15N3
InChi Key:
BQLXMVISWVMAID-UHFFFAOYSA-N
SMILES:
CNCC1=CC=C(CN2C=CC=N2)C=C1