accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 89284-20-8

IUPAC Name: 3-chloro-1,4-dihydropyridin-4-one
Synonyms: 3-chloro-1H-pyridin-4-one
Molecular Weight (g/mol): 129.54
Molecular Formula: C5H4ClNO
InChi Key: NKEYLLFRPHHBOQ-UHFFFAOYSA-N
SMILES: ClC1=CNC=CC1=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results