CAS RN 892845-64-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
2-Chloro-4-fluorobenzeneboronic acid pinacol ester, 96%, Thermo Scientific™
CAS: 892845-64-6 Synonym: 2-2-chloro-4-fluorophenyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane,2-2-chloro-4-fluoro-phenyl-4,4,5,5-tetramethyl-1,3,2 dioxaborolane