accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 893413-42-8

IUPAC Name: [(6-chloro-1H-1,2,3-benzotriazol-1-yl)oxy]tris(pyrrolidin-1-yl)phosphanium; hexafluoro-λ⁵-phosphanuide
Synonyms: [(6-chloro-1,2,3-benzotriazol-1-yl)oxy]tris(pyrrolidin-1-yl)phosphanium hexafluorophosphate
Molecular Weight (g/mol): 554.84
Molecular Formula: C18H27ClF6N6OP2
InChi Key: QJZCQEPNBRAYQL-UHFFFAOYSA-N
SMILES: F[P-](F)(F)(F)(F)F.ClC1=CC=C2N=NN(O[P+](N3CCCC3)(N3CCCC3)N3CCCC3)C2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results