Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(6-Chloro-1H-benzotriazol-1-yloxy)tripyrrolidinophosphonium Hexafluorophosphate 98.0+%, TCI America™
SDP

Catalog No. C34615G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C34615G 5 g
C346125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C34615G Supplier TCI America Supplier No. C34615G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 893413-42-8
Molecular Formula C18H27ClF6N6OP2
Molecular Weight (g/mol) 554.84
MDL Number MFCD14560589
InChI Key QJZCQEPNBRAYQL-UHFFFAOYSA-N
Synonym TPTDP
PubChem CID 16039314
IUPAC Name [(6-chloro-1H-1,2,3-benzotriazol-1-yl)oxy]tris(pyrrolidin-1-yl)phosphanium; hexafluoro-λ⁵-phosphanuide
SMILES F[P-](F)(F)(F)(F)F.ClC1=CC=C2N=NN(O[P+](N3CCCC3)(N3CCCC3)N3CCCC3)C2=C1

Specifications

Melting Point 173°C
Color White-Yellow
Quantity 5 g
Formula Weight 554.84
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material (6-Chloro-1H-benzotriazol-1-yloxy)tripyrrolidinophosphonium Hexafluorophosphate
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.