accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 893556-85-9

IUPAC Name: 10-{[4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-4-ethyl-11-methoxy-3a,7,9,11,13,15-hexamethyl-1-{1-[1-(1,8-naphthyridin-4-yl)ethyl]azetidin-3-yl}-tetradecahydro-1H-oxacyclotetradeca[4,3-d][1,3]oxazole-2,6,8,14-tetrone
Synonyms: 10-{[4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-4-ethyl-11-methoxy-3a,7,9,11,13,15-hexamethyl-1-{1-[1-(1,8-naphthyridin-4-yl)ethyl]azetidin-3-yl}-octahydrooxacyclotetradeca[4,3-d][1,3]oxazole-2,6,8,14-tetrone
Molecular Weight (g/mol): 824.03
Molecular Formula: C44H65N5O10
InChi Key: FATPJRUUPNLGGR-UHFFFAOYNA-N
SMILES: CCC1OC(=O)C(C)C(=O)C(C)C(OC2OC(C)CC(C2O)N(C)C)C(C)(CC(C)C(=O)C(C)C2N(C3CN(C3)C(C)C3=CC=NC4=NC=CC=C34)C(=O)OC12C)OC

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results