Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

PF 945863, Tocris Bioscience™
SDP

Catalog No. 441910 Shop All Novus Biologicals Products
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
44-191-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 44-191-0 Supplier R&D Systems Supplier No. 4419/10

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Macrolide antibiotic

Macrolide antibiotic. Active against multidrug resistant respiratory tract bacterial strains. Does not have the hepatotoxic side effects associated with telithromycin. Orally active.

Chemical Identifiers

CAS 893556-85-9
Molecular Formula C44H65N5O10
Molecular Weight (g/mol) 824.029
InChI Key FATPJRUUPNLGGR-ACVHMLGSSA-N
Synonym 3as,4r,7r,9r,10r,11r,13r,15r,15ar-4-ethyloctahydro-11-methoxy-3a,7,9,11,13,15-hexamethyl-1-1-1r-1-1,8-naphthyridin-4-yl ethyl-3-azetidinyl-10-3,4,6-trideoxy-3-dimethylamino-?-d-xylo-hexopyranosyl oxy-2h-oxacyclotetradecino 4,3-d oxazole-2,6,8,14 1h,7h,9h tetrone
PubChem CID 90488939
IUPAC Name (2R,4R,6R,7R,8R,10R,13R,14S)-7-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-13-ethyl-6-methoxy-2,4,6,8,10,14-hexamethyl-17-[1-[(1R)-1-(1,8-naphthyridin-4-yl)ethyl]azetidin-3-yl]-12,15-dioxa-17-azabicyclo[12.3.0]heptadecane-3,9,11,16-te
SMILES CCC1C2(C(C(C(=O)C(CC(C(C(C(=O)C(C(=O)O1)C)C)OC3C(C(CC(O3)C)N(C)C)O)(C)OC)C)C)N(C(=O)O2)C4CN(C4)C(C)C5=C6C=CC=NC6=NC=C5)C

Specifications

Quantity 10 mg
Formula Weight 824.01
Percent Purity >98%
Chemical Name or Material PF 945863
Recommended Storage Store at -20°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.