accessibility menu, dialog, popup

UserName

Viewing profile as user:
FClNF

CAS RN 89402-43-7

IUPAC Name: 5-chloro-2,3-difluoropyridine
Synonyms: 5-chloro-2,3-difluoropyridine
Molecular Weight (g/mol): 149.52
Molecular Formula: C5H2ClF2N
InChi Key: PERMDYZFNQIKBL-UHFFFAOYSA-N
SMILES: FC1=CC(Cl)=CN=C1F
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results