Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2,3-difluoropyridine 93.0+%, TCI America™
SDP

Catalog No. C21135G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C21135G 5 g
C211325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C21135G Supplier TCI America Supplier No. C21135G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 89402-43-7 | C5H2ClF2N | 149.52 g/mol

Chemical Identifiers

CAS 89402-43-7
Molecular Formula C5H2ClF2N
Molecular Weight (g/mol) 149.52
MDL Number MFCD04039314
InChI Key PERMDYZFNQIKBL-UHFFFAOYSA-N
Synonym 2,3-difluoro-5-chloropyridine, 3-chloro-5,6-difluoropyridine, 5-chloro-2,3-difluoro-pyridine, cdfp, pubchem5411, tpc-i006, 2,3-difluoro-5-chloropyridine;, 5-chloro-2,3-difluoropyridine;, 5-chloro-2,3-difluoro-pyridine;
PubChem CID 2783248
IUPAC Name 5-chloro-2,3-difluoropyridine
SMILES FC1=CC(Cl)=CN=C1F

Specifications

Color Yellow
Boiling Point 135°C
Quantity 5 g
UN Number 1993
Formula Weight 149.52
Percent Purity ≥93.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Chloro-2,3-difluoropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.