CAS RN 89453-69-0
CAS RN 89453-69-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MP Biomedicals, Inc Fast Red TR Salt, MP Biomedicals
CAS: 89453-69-0 Molecular Formula: C14H12Cl6N4Zn Molecular Weight (g/mol): 514.358 InChI Key: RIIVFDKBXXMYRL-UHFFFAOYSA-J PubChem CID: 131675365 IUPAC Name: zinc;4-chloro-2-methylbenzenediazonium;tetrachloride SMILES: CC1=C(C=CC(=C1)Cl)[N+]#N.CC1=C(C=CC(=C1)Cl)[N+]#N.[Cl-].[Cl-].[Cl-].[Cl-].[Zn+2]