Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fast Red TR Salt, MP Biomedicals

Catalog No. ICN19119005 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19119005 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19119005 Supplier MP Biomedicals, Inc Supplier No. 0219119005

Chemical Identifiers

CAS 89453-69-0
Molecular Formula C14H12Cl6N4Zn
Molecular Weight (g/mol) 514.358
InChI Key RIIVFDKBXXMYRL-UHFFFAOYSA-J
PubChem CID 131675365
IUPAC Name zinc;4-chloro-2-methylbenzenediazonium;tetrachloride
SMILES CC1=C(C=CC(=C1)Cl)[N+]#N.CC1=C(C=CC(=C1)Cl)[N+]#N.[Cl-].[Cl-].[Cl-].[Cl-].[Zn+2]

Specifications

Assay Percent Range Min. 90%
Formula Weight 257.2
Quantity 5 g
Chemical Name or Material Fast Red TR Salt, Zinc Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.