CAS RN 89535-60-4
CAS RN 89535-60-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Poly(1,1-dimethylsilazane), telomer, Thermo Scientific™
CAS: 89535-60-4 Molecular Formula: [-Si(CH3)2NH-]n MDL Number: MFCD00241406