CAS RN 896138-40-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
MilliporeSigma™ Calbiochem™ Flt-3 Inhibitor II
The Flt-3 Inhibitor II, also referenced under CAS 896138-40-2, controls the biological activity of Flt-3.