Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Calbiochem™ Flt-3 Inhibitor II

Catalog No. 3430211MG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
34-302-11MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 34-302-11MG Supplier MilliporeSigma™ Supplier No. 3430211MG

missing translation for 'validMainframeMsg'

The Flt-3 Inhibitor II, also referenced under CAS 896138-40-2, controls the biological activity of Flt-3.

  • Packaged Under Inert Gas
  • Cell Permeable
TRUSTED_SUSTAINABILITY

Specifications

Color Yellowish Brown
Description 95% by HPLC
Target Proteins Flt-3
CAS 896138-40-2
Molecular Formula C17H12N2O3
Formula Weight 292.3g/mol
Solubility DMSO or EtOH
Content And Storage Protect from light
Form Solid
Quantity 1 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.