accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3NH3CN(E)

CAS RN 897-55-2

IUPAC Name: N,N-dimethyl-4-[(1E)-2-(quinolin-4-yl)ethenyl]aniline
Synonyms: N,N-dimethyl-4-[(1E)-2-(quinolin-4-yl)ethenyl]aniline
Molecular Weight (g/mol): 274.37
Molecular Formula: C19H18N2
InChi Key: CIXDQQGMRYRUQA-JXMROGBWSA-N
SMILES: CN(C)C1=CC=C(\C=C\C2=C3C=CC=CC3=NC=C2)C=C1
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results