Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

4-(4-Dimethylaminostyryl)quinoline 96.0+%, TCI America™
SDP

Catalog No. D22761G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22761G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D22761G Supplier TCI America Supplier No. D22761G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 897-55-2
Molecular Formula C19H18N2
Molecular Weight (g/mol) 274.367
MDL Number MFCD00191349
InChI Key CIXDQQGMRYRUQA-YFHOEESVSA-N
PubChem CID 1550830
IUPAC Name N,N-dimethyl-4-[(Z)-2-quinolin-4-ylethenyl]aniline
SMILES CN(C)C1=CC=C(C=C1)C=CC2=CC=NC3=CC=CC=C23

Specifications

Melting Point 140°C
Quantity 1 g
Formula Weight 274.37
Percent Purity ≥96.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-(4-Dimethylaminostyryl)quinoline
RTECSNumber VB7000000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.