accessibility menu, dialog, popup

UserName

Viewing profile as user:
CH3CH3H3COONHO

CAS RN 89711-08-0

IUPAC Name: tert-butyl N-(2-oxoethyl)carbamate
Synonyms: tert-butyl N-(2-oxoethyl)carbamate
Molecular Weight (g/mol): 159.19
Molecular Formula: C7H13NO3
InChi Key: ACNRTYKOPZDRCO-UHFFFAOYSA-N
SMILES: CC(C)(C)OC(=O)NCC=O
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results