CAS RN 89778-26-7
IUPAC Name:
(2-{4-[(1Z)-4-chloro-1,2-diphenylbut-1-en-1-yl]phenoxy}ethyl)dimethylamine
Synonyms:
toremifene|fareston|acapodene|farestone|citrate, toremifene|Z-toremifene|toremifeno|toremifene base|TOR|toremifenum
Molecular Weight (g/mol):
405.97
Molecular Formula:
C26H28ClNO
InChi Key:
XFCLJVABOIYOMF-QPLCGJKRSA-N
SMILES:
CN(C)CCOC1=CC=C(C=C1)C(=C(\CCCl)C1=CC=CC=C1)\C1=CC=CC=C1
1
–
1
of
1
results
1
Thermo Scientific Chemicals Toremifene, 98+%
CAS: 89778-26-7 Molecular Formula: C26H28ClNO,C26H28ClNO Synonym: Toremifene base; 2-(4-[(1Z)-4-Chloro-1,2-diphenyl-1-butenyl]phenoxy)-N,N-dimethylethanamine