accessibility menu, dialog, popup

UserName

Viewing profile as user:

CAS RN 89786-04-9

IUPAC Name: (2S,3S,5R)-3-methyl-4,4,7-trioxo-3-[(1H-1,2,3-triazol-1-yl)methyl]-4λ⁶-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
Synonyms: tazobactam
Molecular Weight (g/mol): 300.29
Molecular Formula: C10H12N4O5S
InChi Key: LPQZKKCYTLCDGQ-WEDXCCLWSA-N
SMILES: C[C@]1(CN2C=CN=N2)[C@@H](N2[C@@H](CC2=O)S1(=O)=O)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results