Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tazobactam, 94%

Catalog No. AAJ6648303
Change view
Click to view available options
Quantity:
1 g
Packaging:
Plastic bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AAJ6648303 1 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ6648303 Supplier Thermo Scientific Chemicals Supplier No. J6648303
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 89786-04-9 | C10H12N4O5S | 300.29 g/mol

Tazobactam is a drug is used in conjunction with beta-lactamase susceptible penicillins to treat infections caused by beta-lactamase producing organisms. It acts as a beta lactamase inhibitor. It is used in combination with piperacillin to broaden the spectrum of piperacillin antibacterial action. It is used as antibiotic in the treatment of nosocomial pneumonia caused by Pseudomonas aeruginosa.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89786-04-9
Molecular Formula C10H12N4O5S
Molecular Weight (g/mol) 300.29
MDL Number MFCD00917472 MFCD00867002
InChI Key LPQZKKCYTLCDGQ-WEDXCCLWSA-N
Synonym tazobactam, tazobactam acid, tazobactamum, zosyn, unii-se10g96m8w, ytr-830h, ytr 830h, chembl404, ytr830h, 2s,3s,5r-3-methyl-7-oxo-3-1h-1,2,3-triazol-1-ylmethyl-4-thia-1-azabicyclo 3.2.0 heptane-2-carboxylic acid 4,4-dioxide
PubChem CID 123630
ChEBI CHEBI:9421
IUPAC Name (2S,3S,5R)-3-methyl-4,4,7-trioxo-3-[(1H-1,2,3-triazol-1-yl)methyl]-4λ⁶-thia-1-azabicyclo[3.2.0]heptane-2-carboxylic acid
SMILES C[C@]1(CN2C=CN=N2)[C@@H](N2[C@@H](CC2=O)S1(=O)=O)C(O)=O

Specifications

Melting Point 115°C to 145°C
Boiling Point 77°C
Flash Point >110°C (230°F)
Packaging Plastic bottle
Quantity 1 g
Solubility Information Soluble in water
Formula Weight 300.29
Percent Purity 94%
Physical Form Powder
Chemical Name or Material Tazobactam
Hazard Category H317-H334
Precautionary Statement P261-P272-P280g-P285-P302+P352-P304+P341-P333+P313-P342+P311-P363-P501c
RTECSNumber XI0191400
TSCA No
Recommended Storage Store at -20°C

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.